accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1423031-35-9

IUPAC Name: imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Synonyms: imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Molecular Weight (g/mol): 198.61
Molecular Formula: C8H7ClN2O2
InChi Key: OQGRMPZJJDBYBL-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CC2=NC=CN2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results