accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FHNSHNCl

CAS RN 142313-30-2

IUPAC Name: 5-chloro-6-fluoro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5-chloro-6-fluoro-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 202.63
Molecular Formula: C7H4ClFN2S
InChi Key: DQXCFZSRGLNXKW-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C2NC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results