accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

AuPClPhPhPh

CAS RN 14243-64-2

IUPAC Name: gold(1+) triphenylphosphane chloride
Synonyms: chloro(triphenylphosphine)gold(I)
Molecular Weight (g/mol): 494.71
Molecular Formula: C18H15AuClP
InChi Key: IFPWCRBNZXUWGC-UHFFFAOYSA-M
SMILES: [Cl-].[Au+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)