accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOFFOCH3

CAS RN 142492-01-1

IUPAC Name: 2,2-difluoro-N-methoxy-N-methylacetamide
Synonyms: 2,2-difluoro-N-methoxy-N-methylacetamide
Molecular Weight (g/mol): 139.1
Molecular Formula: C4H7F2NO2
InChi Key: CUPRFYMJGQMIIC-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C(F)F
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)