accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14263-94-6

IUPAC Name: zinc(2+) 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-bis(diazonium) tetrachloride
Synonyms: zinc(2+) 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-bis(diazonium) tetrachloride
Molecular Weight (g/mol): 475.46
Molecular Formula: C14H12Cl4N4O2Zn
InChi Key: GPPKNJIWDULNQH-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Zn++].COC1=CC(=CC=C1[N+]#N)C1=CC=C([N+]#N)C(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results