accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14264-16-5

IUPAC Name: nickel(2+) bis(triphenylphosphane) dichloride
Synonyms: nickel(2+) bis(triphenylphosphine) dichloride
Molecular Weight (g/mol): 654.18
Molecular Formula: C36H30Cl2NiP2
InChi Key: ZBRJXVVKPBZPAN-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ni++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results