accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OOSOOOOSOOOOSOOOFeFe

CAS RN 142906-29-4

IUPAC Name: diiron(3+) pentahydrate trisulfate
Synonyms: diiron(3+) pentahydrate trisulfate
Molecular Weight (g/mol): 489.93
Molecular Formula: Fe2H10O17S3
InChi Key: YHGPYBQVSJBGHH-UHFFFAOYSA-H
SMILES: O.O.O.O.O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)