CAS RN 14292-29-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxyhexanedioic Acid, TRC
CAS: 14292-29-6 Molecular Formula: C6 H10 O5 Molecular Weight (g/mol): 162.14 Synonym: Hexanedioic acid, 3-hydroxy-,3-Hydroxyhexanedioic acid,3-Hydroxyadipic acid,β-Hydroxyadipic acid IUPAC Name: 3-hydroxyhexanedioic acid SMILES: OC(CCC(=O)O)CC(=O)O