accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NSNO

CAS RN 14294-10-1

IUPAC Name: morpholine-4-carbothioamide
Synonyms: morpholine-4-carbothioamide
Molecular Weight (g/mol): 146.21
Molecular Formula: C5H10N2OS
InChi Key: GSLBUBZXFUYMSW-UHFFFAOYSA-N
SMILES: NC(=S)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results