accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOClFFF

CAS RN 142994-09-0

IUPAC Name: 4-chloro-2-(trifluoromethyl)benzoic acid
Synonyms: 4-chloro-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: RKZXXMAQKMOZLK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results