accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSH

CAS RN 143-10-2

IUPAC Name: decane-1-thiol
Synonyms: decane-1-thiol
Molecular Weight (g/mol): 174.35
Molecular Formula: C10H22S
InChi Key: VTXVGVNLYGSIAR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCS
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results