accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14305-08-9

IUPAC Name: 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one
Synonyms: 4,5-dibromo-2-phenylpyridazin-3-one
Molecular Weight (g/mol): 329.98
Molecular Formula: C10H6Br2N2O
InChi Key: NQJXEMRMTOSSBQ-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(=O)N(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results