accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 143158-15-0

OHFFFFFF

CAS RN 143158-15-0

IUPAC Name: [2,4-bis(trifluoromethyl)phenyl]methanol
Synonyms: [2,4-bis(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 244.14
Molecular Formula: C9H6F6O
InChi Key: NDHDRXKQSOFDNV-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results