accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14331-56-7

IUPAC Name: 4-hydrazinyl-2,6-dimethylpyrimidine
Synonyms: 4-hydrazinyl-2,6-dimethylpyrimidine
Molecular Weight (g/mol): 138.17
Molecular Formula: C6H10N4
InChi Key: HVAYIAWJHRUOBC-UHFFFAOYSA-N
SMILES: CC1=CC(NN)=NC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results