accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BFFFFCH3NNH3C

CAS RN 143314-16-3

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1-ethyl-3-methylimidazolium tetrafluoroborate
Molecular Weight (g/mol): 197.97
Molecular Formula: C6H11BF4N2
InChi Key: CUNYTRQQXKCRTJ-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCN1C=C[N+](C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results