accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH3CCH3OO

CAS RN 143314-17-4

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium acetate
Synonyms: 1-ethyl-3-methylimidazolium acetate
Molecular Weight (g/mol): 170.21
Molecular Formula: C8H14N2O2
InChi Key: XIYUIMLQTKODPS-UHFFFAOYSA-M
SMILES: CC([O-])=O.CCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results