CAS RN 143322-58-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Eletriptan, TRC
CAS: 143322-58-1 Molecular Formula: C22 H26 N2 O2 S Molecular Weight (g/mol): 382.52 Synonym: 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole,1H-Indole, 3-[(1-methyl-2-pyrrolidinyl)methyl]-5-[2-(phenylsulfonyl)ethyl]-, (R)-,(R)-5-[2-(Benzenesulfonyl)ethyl]-3-[(N-methylpyrrolidin-2-yl)methyl]-1H-indole,Eletriptan,UK 116044 IUPAC Name: 5-[2-(benzenesulfonyl)ethyl]-3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]-1H-indole SMILES: CN1CCC[C@@H]1Cc2c[nH]c3ccc(CCS(=O)(=O)c4ccccc4)cc23