accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1435-44-5

FClF

CAS RN 1435-44-5

IUPAC Name: 1-chloro-2,4-difluorobenzene
Synonyms: 1-chloro-2,4-difluorobenzene
Molecular Weight (g/mol): 148.54
Molecular Formula: C6H3ClF2
InChi Key: AJCSNHQKXUSMMY-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results