accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NN

CAS RN 1436-43-7

IUPAC Name: quinoline-2-carbonitrile
Synonyms: 2-cyanoquinoline
Molecular Weight (g/mol): 154.17
Molecular Formula: C10H6N2
InChi Key: WDXARTMCIRVMAE-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2C=CC=CC2=N1
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results