CAS RN 144-62-7
Oxalic acid, 98%, anhydrous
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, anhydrous, 98%
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, 10% w/v aq. soln.
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, 5% w/v aq. soln.
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, 0.1 N standard solution
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 MDL Number: MFCD00002573 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid solution, Volumetric, 0.5 M (COOH)2 (1N), Solstice
CAS: 144-62-7 Molecular Formula: C2H2O4 Molecular Weight (g/mol): 90.034 InChI Key: MUBZPKHOEPUJKR-UHFFFAOYSA-N Synonym: ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico PubChem CID: 971 ChEBI: CHEBI:16995 IUPAC Name: oxalic acid SMILES: C(=O)(C(=O)O)O
Oxalic acid, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Oxalic Acid, TRC
CAS: 144-62-7 Molecular Formula: C2 H2 O4 Molecular Weight (g/mol): 90.03 Synonym: Oxalic Acid,Ethanedioic acid,Oxalic acid,Aktisal,Aquisal,DeerClean,NSC 132055,NSC 151956,NSC 62774,NSC 76990,Ultraplast Activate S 52 IUPAC Name: oxalic acid SMILES: OC(=O)C(=O)O
Oxalic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Oxalic Acid Solution, 0.05M, Solstice
- 1
- 2