accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 144025-03-6

IUPAC Name: (2,4-difluorophenyl)boronic acid
Synonyms: 2,4-difluorophenylboronic acid
Molecular Weight (g/mol): 157.91
Molecular Formula: C6H5BF2O2
InChi Key: QQLRSCZSKQTFGY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C=C1F
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results