accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14414-32-5

H3COHNNHOCH3OOH3COH3CO

CAS RN 14414-32-5

IUPAC Name: 4-({2-[(3,5-dimethoxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]hydrazin-1-yl}methylidene)-2,6-dimethoxycyclohexa-2,5-dien-1-one
Synonyms: 4-({2-[(3,5-dimethoxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]hydrazin-1-yl}methylidene)-2,6-dimethoxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 360.37
Molecular Formula: C18H20N2O6
InChi Key: MYNVJNZGTAVRJD-UHFFFAOYSA-N
SMILES: COC1=CC(=CNNC=C2C=C(OC)C(=O)C(OC)=C2)C=C(OC)C1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)