CAS RN 144143-96-4
IUPAC Name:
4-({2-butyl-5-[(1E)-2-carboxy-2-[(thiophen-2-yl)methyl]eth-1-en-1-yl]-1H-imidazol-1-yl}methyl)benzoic acid; methanesulfonic acid
Synonyms:
teveten eprosartan mesylate eprosartan methanesulfonate
Molecular Weight (g/mol):
520.62
Molecular Formula:
C24H28N2O7S2
InChi Key:
DJSLTDBPKHORNY-XMMWENQYSA-N
SMILES:
CS(O)(=O)=O.CCCCC1=NC=C(\C=C(/CC2=CC=CS2)C(O)=O)N1CC1=CC=C(C=C1)C(O)=O
1
–
3
of
3
results
1
Eprosartan Mesilate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Eprosartan Mesylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.