accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 144222-22-0

CH3H3CCH3OONNH2

CAS RN 144222-22-0

IUPAC Name: {1-[(tert-butoxy)carbonyl]piperidin-4-yl}methanaminium
Synonyms: [1-(tert-butoxycarbonyl)piperidin-4-yl]methanaminium
Molecular Weight (g/mol): 215.32
Molecular Formula: C11H23N2O2
InChi Key: KLKBCNDBOVRQIJ-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)N1CCC(C[NH3+])CC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)