CAS RN 14434-22-1
CAS RN 14434-22-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
484.06
Molecular Formula:
C6H20FeN6Na4O10
InChi Key:
JFSUDVTVQZUDOP-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
1
–
2
of
2
results
1
Sodium ferrocyanide decahydrate
CAS: 14434-22-1 Molecular Formula: C6H20FeN6Na4O10 Molecular Weight (g/mol): 484.06 MDL Number: MFCD00149274 InChI Key: JFSUDVTVQZUDOP-UHFFFAOYSA-N SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
2
Sodium hexacyanoferrate(II) decahydrate, 99%
CAS: 14434-22-1 Molecular Formula: C6H20FeN6Na4O10 Molecular Weight (g/mol): 484.06 MDL Number: MFCD00149274 InChI Key: JFSUDVTVQZUDOP-UHFFFAOYSA-N Synonym: Sodium ferrocyanide decahydrate SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N