accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14438-32-5

IUPAC Name: 2-amino-3,5-dimethylbenzoic acid
Synonyms: 2-amino-3,5-dimethylbenzoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: GIMYRAQQQBFFFJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N)C(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results