accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHFCl

CAS RN 144432-85-9

IUPAC Name: (3-chloro-4-fluorophenyl)boronic acid
Synonyms: 3-chloro-4-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: WJDZZXIDQYKVDG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C(Cl)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results