accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14444-77-0

IUPAC Name: (diethoxymethoxy)benzene
Synonyms: (diethoxymethoxy)benzene
Molecular Weight (g/mol): 196.25
Molecular Formula: C11H16O3
InChi Key: QTURWMMVIIBRRP-UHFFFAOYSA-N
SMILES: CCOC(OCC)OC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results