accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO(R)O(S)(R)HO(S)HONHOHNO

CAS RN 1445-07-4

IUPAC Name: 5-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: β-pseudouridine β-D-pseudouridine pseudouridine 5-β-delta-ribofuranosyl-uracil β-delta-pseudouridine y-uridine pseudouridine C psi-uridine
Molecular Weight (g/mol): 244.2
Molecular Formula: C9H12N2O6
InChi Key: PTJWIQPHWPFNBW-GBNDHIKLSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)C1=CNC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)