accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14457-87-5

BrBrBrCe

CAS RN 14457-87-5

IUPAC Name: cerium(3+) tribromide
Synonyms: cerium(III) bromide
Molecular Weight (g/mol): 379.83
Molecular Formula: Br3Ce
InChi Key: MOOUSOJAOQPDEH-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Ce+3]
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)