CAS RN 1446144-71-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acetaminotadalafil, TRC
CAS: 1446144-71-3 Molecular Formula: C23 H20 N4 O5 Molecular Weight (g/mol): 432.43 Synonym: Acetamide, N-[(6R,12aR)-6-(1,3-benzodioxol-5-yl)-3,4,6,7,12,12a-hexahydro-1,4-dioxopyrazino[1',2':1,6]pyrido[3,4-b]indol-2(1H)-yl]-,N-[(6R,12aR)-6-(1,3-Benzodioxol-5-yl)-3,4,6,7,12,12a-hexahydro-1,4-dioxopyrazino[1',2':1,6]pyrido[3,4-b]indol-2(1H)-yl]acetamide,Acetaminotadalafil IUPAC Name: N-[(2R,8R)-2-(1,3-benzodioxol-5-yl)-4,7-dioxo-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-6-yl]acetamide SMILES: CC(=O)NN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O