accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHNOOCH3OOCH3

CAS RN 144689-94-1

IUPAC Name: 4,5-diethyl 2-propyl-1H-imidazole-4,5-dicarboxylate
Synonyms: 4,5-diethyl 2-propyl-1H-imidazole-4,5-dicarboxylate
Molecular Weight (g/mol): 254.29
Molecular Formula: C12H18N2O4
InChi Key: ZQPMIDPIXSMOAD-UHFFFAOYSA-N
SMILES: CCCC1=NC(C(=O)OCC)=C(N1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results