accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCl

CAS RN 14470-28-1

IUPAC Name: 1-(chlorodiphenylmethyl)-4-methoxybenzene
Synonyms: 1-(chlorodiphenylmethyl)-4-methoxybenzene
Molecular Weight (g/mol): 308.81
Molecular Formula: C20H17ClO
InChi Key: OBOHMJWDFPBPKD-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)