accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FSFFFFFOOH

CAS RN 1448319-13-8

IUPAC Name: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 266.16
Molecular Formula: C7H4F6O2S
InChi Key: WQRFQFPUMUJGOL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results