accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 144851-62-7

IUPAC Name: 2,6-difluoro-3-methoxyaniline
Synonyms: 2,6-difluoro-3-methoxyaniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: XVLHUQRLEPQGDI-UHFFFAOYSA-N
SMILES: COC1=C(F)C(N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results