accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1449133-71-4

IUPAC Name: (2-chloro-5-{[(3-fluorophenyl)methyl]carbamoyl}phenyl)boronic acid
Synonyms: 2-chloro-5-{[(3-fluorophenyl)methyl]carbamoyl}phenylboronic acid
Molecular Weight (g/mol): 307.51
Molecular Formula: C14H12BClFNO3
InChi Key: BULLMNJRUBFQST-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(=C1)C(=O)NCC1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results