accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 144918-48-9

IUPAC Name: 4-methyl-N-[3-(trifluoromethyl)phenyl]benzamide
Synonyms: 4-methyl-N-[3-(trifluoromethyl)phenyl]benzamide
Molecular Weight (g/mol): 279.26
Molecular Formula: C15H12F3NO
InChi Key: DSGGHRHTHCRAAW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NC1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results