accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOH3COOCH3OOCH3

CAS RN 14495-41-1

IUPAC Name: 1,2,3,4-tetramethyl cyclobutane-1,2,3,4-tetracarboxylate
Synonyms: 1,2,3,4-tetramethyl cyclobutane-1,2,3,4-tetracarboxylate
Molecular Weight (g/mol): 288.25
Molecular Formula: C12H16O8
InChi Key: NXMOMNBIDWYNOP-UHFFFAOYSA-N
SMILES: COC(=O)C1C(C(C1C(=O)OC)C(=O)OC)C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results