accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 145-50-6

IUPAC Name: (4E)-4-[(4-hydroxynaphthalen-1-yl)(phenyl)methylidene]-1,4-dihydronaphthalen-1-one
Synonyms: (4E)-4-[(4-hydroxynaphthalen-1-yl)(phenyl)methylidene]naphthalen-1-one
Molecular Weight (g/mol): 374.44
Molecular Formula: C27H18O2
InChi Key: VDDWRTZCUJCDJM-SOYKGTTHSA-N
SMILES: OC1=C2C=CC=CC2=C(C=C1)C(\C1=CC=CC=C1)=C1/C=CC(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)