accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHSOOCl

CAS RN 145-74-4

IUPAC Name: 8-chloronaphthalene-1-sulfonic acid
Synonyms: 8-chloronaphthalene-1-sulfonic acid
Molecular Weight (g/mol): 242.67
Molecular Formula: C10H7ClO3S
InChi Key: OPFNCYFEIBUZHU-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=C2C(Cl)=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results