accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1450-14-2

IUPAC Name: hexamethyldisilane
Synonyms: hexamethyldisilane
Molecular Weight (g/mol): 146.38
Molecular Formula: C6H18Si2
InChi Key: NEXSMEBSBIABKL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)[Si](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results