accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONOO

CAS RN 1450-76-6

IUPAC Name: 2-acetyl-4-nitrobenzen-1-olate
Synonyms: 2-acetyl-4-nitrobenzenolate
Molecular Weight (g/mol): 180.14
Molecular Formula: C8H6NO4
InChi Key: LNCBPUWMGYOISS-UHFFFAOYSA-M
SMILES: CC(=O)C1=CC(=CC=C1[O-])[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results