accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14508-49-7

IUPAC Name: 2-chloropyrazine
Synonyms: 2-chloropyrazine
Molecular Weight (g/mol): 114.53
Molecular Formula: C4H3ClN2
InChi Key: GELVZYOEQVJIRR-UHFFFAOYSA-N
SMILES: ClC1=CN=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results