accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1451-82-7

CH3OCH3Br

CAS RN 1451-82-7

IUPAC Name: 2-bromo-1-(4-methylphenyl)propan-1-one
Synonyms: 2-bromo-1-(4-methylphenyl)propan-1-one
Molecular Weight (g/mol): 227.1
Molecular Formula: C10H11BrO
InChi Key: OZLUPIIIHOOPNQ-UHFFFAOYNA-N
SMILES: CC(Br)C(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)