accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONH2NHClHCl

CAS RN 145148-39-6

IUPAC Name: 1-[(2-methoxyphenyl)methyl]-2-phenylpiperidin-3-amine dihydrochloride
Synonyms: 1-[(2-methoxyphenyl)methyl]-2-phenylpiperidin-3-amine dihydrochloride
Molecular Weight (g/mol): 369.33
Molecular Formula: C19H26Cl2N2O
InChi Key: STUCSMJJXNCIOE-UHFFFAOYNA-N
SMILES: Cl.Cl.COC1=CC=CC=C1CN1CCCC(N)C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results