accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

COCOCOCOCOMnBr

CAS RN 14516-54-2

IUPAC Name: pentakis(methanidylidyneoxidanium) manganese bromide
Synonyms: pentakis(carbon monoxide) manganese bromide
Molecular Weight (g/mol): 274.89
Molecular Formula: C5BrMnO5
InChi Key: LOCZKKJCQVRJPQ-UHFFFAOYSA-M
SMILES: [Mn].[Br-].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results