accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(R)(R)(S)(R)(S)(S)(R)(S)(R)(R)OHOHHHHHOH2ONaOHMeMeMe

CAS RN 145224-92-6

IUPAC Name: 4-{7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoic acid hydrate sodium
Synonyms: acid, deoxycholic hydrate sodium
Molecular Weight (g/mol): 433.59
Molecular Formula: C24H42NaO5
InChi Key: HKLBTQUQYACMJN-UHFFFAOYNA-N
SMILES: O.[Na].CC(CCC(O)=O)C1CCC2C3CCC4CC(O)CCC4(C)C3CC(O)C12C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results