accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1454-53-1

CH3OONOHCl

CAS RN 1454-53-1

IUPAC Name: ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloridyl
Synonyms: ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloridyl
Molecular Weight (g/mol): 296.77
Molecular Formula: C15H19ClNO3
InChi Key: RHLGESNVVDXRQP-UHFFFAOYNA-N
SMILES: [Cl].CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)