CAS RN 1454889-37-2
CAS RN 1454889-37-2
IUPAC Name:
({5-[6-(methylamino)-9H-purin-9-yl]-2-[(phosphonooxy)methyl]oxolan-3-yl}oxy)phosphonic acid tetrasodium
Synonyms:
{5-[6-(methylamino)purin-9-yl]-2-[(phosphonooxy)methyl]oxolan-3-yl}oxyphosphonic acid tetrasodium
Molecular Weight (g/mol):
517.19
Molecular Formula:
C11H17N5Na4O9P2
InChi Key:
HLJPLHFSEAJJGC-UHFFFAOYNA-N
SMILES:
[Na].[Na].[Na].[Na].CNC1=C2N=CN(C3CC(OP(O)(O)=O)C(COP(O)(O)=O)O3)C2=NC=N1
1
–
1
of
1
results
1
Tocris Bioscience™ MRS 2179 tetrasodium salt
CAS: 1454889-37-2 Molecular Formula: C11H17N5Na4O9P2