accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14553-08-3

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHEr

CAS RN 14553-08-3

IUPAC Name: tris(4-hydroxypent-3-en-2-one) erbium
Synonyms: tris(4-hydroxypent-3-en-2-one) erbium
Molecular Weight (g/mol): 467.61
Molecular Formula: C15H24ErO6
InChi Key: MUXSKGOXFIMTKO-UHFFFAOYSA-N
SMILES: [Er].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results